missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Reusable Gas/Media Handling Fittings For 6L Angled Sidearm Flasks

Includes PET insert with fluorocarbon O-ring and polypropylene-sealing cap

Brand:  Corning™ 4519-164

Product Code. 10042212

  • 541.35 € / Each
Estimated Shipment: 19-09-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Used to aseptically transfer medium, sparge the cell culture medium directly or add gases to the head space above the cell culture medium
  • Fittings are comprised of a PET insert with a fluorocarbon O-Ring and a polypropylene sealing cap.
  • One or two 316 stainless steel tubes extend to the bottom of the flask; the other is a shorter 6 length.
  • Both tubes are held in place by modified polyphenylene oxide thermoplastic nuts with integrated ferrules.
  • The fittings are completely autoclavable
Spare parts for sidearm fittings, see Cat. Nos. 1395-32LTC, 1395-45LTC, 1395-LTR, and 1395LTMC
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tube style: Angled
1.9 cm
PET insert w/fluorocarbon O-ring and polypropylene-sealing cap
1/Pk.
Non-sterile
1/4 in.
6L Angled Sidearm Flasks
1/Cs.
Media/Gas handling fitting
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Reusable Gas/Media Handling Fittings For 6L Angled Sidearm Flasks > 6 L; 1/4 in.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.