missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PureCoat™ Collagen I Peptide TC-Treated Microplates
GREENER_CHOICE

Product Code. p-4658754
Click to view available options
Well Count:
6
24
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 15300011

Brand: Corning™ 356270

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Lid

Coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface

Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.

Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance cGMP
Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Color Clear
Culture Area 9.6 cm2
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Cell Culture
Type Cell Culture Microplate
Well Count 6
For Use With (Application) Provides a highly consistent alternative to complex extracellular peptides
Includes Lid
No. per Case 10/Cs.
No. per Pack 5/Pk.
Storage Requirements Room Temperature
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.