Learn More
Corning™ PureCoat™ Collagen Peptide Cell Culture Flasks with Vented Cap
Coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface
Brand:  Corning™ 356272
 View more versions of this product
Product Code. 15340011
  • 397.60 € / Pack of 10
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.
Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polystyrene
10/Cs.
Rectangular
Rectangular
75 cm2
Canted
5/Pk.
PureCoat E cm Mimetic Collagen I Peptide
Vented