missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Sterile Spatulas, V-scoop



Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™ Sterile Spatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.
Brand: Corning™ 3006
240.09 EUR valid until 2025-03-31
Use promo code "24073" to get your promotional price.
This item is not returnable.
View return policy
Description
- Corning spatulas are designed to save time and to provide contamination-free samples.
- Eliminates the recycling and resterilizing necessary with reusable spatulas.
- Available in five different configurations.
- Microspatulas available in two configurations.
- Individually packaged.
- Antistatic.
- Sterile.
- RNase-/DNase-free.
- Nonpyrogenic.

Specifications
Sterile | |
Single-use | |
Molecular biology applications, lyophilized samples and contamination-free sampling | |
V-scoop at one end and a larger round spoon at the other end | |
24.9 cm |
USP Class VI, non-pyrogenic, RNase and DNase-free Certified | |
Spatula | |
9.8 in. | |
Polystyrene |
COR001 Lifescience Guid
File size: 7 MB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Sterile Spatulas, V-scoop > Spatula, V-Scoop/Spoon
Spot an opportunity for improvement?Share a Content Correction