Learn More
Corning™ Stripwell™ Microplates
GREENER_CHOICE
Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Microplates provide a superior solution for in vitro diagnostic assays.
Brand:  Corning™ 2593
Product Code. 10367221
  • 623.83 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Strip Plate
8 x 12
Clear
96
Non-sterile
360 μL
In-vitro diagnostic assays
25/Pk.
Without Lid
Medium Binding, Egg crate style
Polystyrene
75 to 200 μL
Non-treated
Round
100/Cs.
100/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Stripwell™ Microplates > Medium; Clear; 25/Pk