Learn More
Corning™ TC-Treated Culture Dishes
BIOPHARMAGREENER_CHOICE
Tissue-culture (TC) treated for optimum attachment and growth of anchorage-dependent cells. Corning™ TC-Treated Culture Dishes provide an ideal solution for growing and maintaining cells in vitro.
240.00 € - 485.00 €
Specifications
Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Color Clear
View More Specs
Products 7
Product Code Brand Shape Culture Area Description Gridded Price Quantity & Availability  
Product Code Brand Shape Culture Area Description Gridded Price Quantity & Availability  
10310272
GREENER_CHOICE
View Documents
Corning™
430196
Round 21 cm2 51.4 dia. x 15 mm H Gridded
359.63 €
Pack of 500
Estimated Shipment: 20-06-2025
to see stock.
Add to basket
10350602
GREENER_CHOICE
View Documents
Corning™
431110
Square 500 cm2 245 L x 245 W x 25 mm H Not Gridded
250.00 €
Pack of 16
In Stock
to see stock.
Edge
Add to basket
10095381
BIOPHARMAGREENER_CHOICE
View Documents
Corning™
430293
Round 55 cm2 80.5 dia. x 20 mm H Not Gridded
472.48 €
Pack of 480
In Stock
to see stock.
Edge
Add to basket
10556661
GREENER_CHOICE
View Documents
Corning™
430165
Round 9 cm2 34.4 dia. x 10 mm H Not Gridded
295.70 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
10634141
GREENER_CHOICE
View Documents
Corning™
430166
Round 21 cm2 52.1 dia. x 15 mm H Not Gridded
323.25 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
10075371
GREENER_CHOICE
View Documents
Corning™
430167
Round 55 cm2 83.8 dia. x 20 mm H Not Gridded
485.00 €
Pack of 500
In Stock
to see stock.
Add to basket
10117320
BIOPHARMAGREENER_CHOICE
View Documents
Corning™
430599
Round 148 cm2 134.5 dia. x 25 mm H Not Gridded
240.00 €
Pack of 60
In Stock
to see stock.
Add to basket
Description
Description
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment
  • Ultra-Low Attachment surface dishes feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation
  • 10 dishes/bag are available for 100 mm dishes (Cat. No. 430293)
  • 245 mm square dishes offer 500 cm2 growth surface
  • Stacking beads aid in handling
  • Vents provide consistent gas exchange
  • Manufactured from optically clear virgin polystyrene
  • Sterile
  • Nonpyrogenic
Dish style (mm) = actual growth surface diameters: 35 mm dish = 34.4 mm; 60 mm dish = 52.1 mm; 100 mm dish = 83.8 mm; 150 mm dish = 139.1 mm.
Cat. no. 431110, Square dishes with interior bottom dimensions of 224 x 224 mm.
Order Info
Package quantities differ. Check ordering table for details.
Specifications
Specifications
Clear
Polystyrene
Clear
Polystyrene
Cell Culture
Lid
Yes
Flat
Single-use
Sterile
Cell Culture Petri Dish