Learn More
Corning™ Thermowell™ 96-Well Polycarbonate PCR Microplates
GREENER_CHOICE
Manufactured from ultrathin polycarbonate for fast, consistent heat transfer. Corning™ Thermowell™ 96-Well Polycarbonate PCR Microplates are ideal for the polyerase chain reaction (PCR) method of DNA amplification.
Brand:  Corning™ 6511
 View more versions of this product
Product Code. 10130961
  • 172.49 € / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Low protein-binding surface prevents loss of sample
  • For oil-free operation, use polyethylene or aluminum sealing tape in conjunction with heated-lid thermal cyclers; also accommodates silicone reusable sealing mat
  • Withstands temperatures to 135°C without warping
  • Various well shapes fit popular thermal cyclers
  • Surface of DNA-BIND™ plate allows covalent attachment of ssDNA at 5« and 3« end
  • Nonsterile
Certifications
Polypropylene plates are certified DNase- and RNase-free.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
8 x 12
Clear
Polycarbonate
Non-sterile
PCR
1/Pk.
Without Lid
Medium Binding
MJ Research PTC-100-96V, PTC-200 DNA EngineTM, Biometra Uno - ThermoblockerTM,Coy Corporation Temp Cycler II, Corbett Research FTS-960, Hybaid OmniGene withMicroblock, Quatro BioSystems T-C-40
96
Round
25/Cs.
Clear
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products