missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Thermowell™ 96-Well Polycarbonate PCR Microplates
Manufactured from ultrathin polycarbonate for fast, consistent heat transfer. Corning™ Thermowell™ 96-Well Polycarbonate PCR Microplates are ideal for the polyerase chain reaction (PCR) method of DNA amplification.
Brand: Corning™ 6509
149.12 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.
This item is not returnable.
View return policy
Description
- Low protein-binding surface prevents loss of sample
- For oil-free operation, use polyethylene or aluminum sealing tape in conjunction with heated-lid thermal cyclers; also accommodates silicone reusable sealing mat
- Withstands temperatures to 135°C without warping
- Various well shapes fit popular thermal cyclers
- Surface of DNA-BIND™ plate allows covalent attachment of ssDNA at 5« and 3« end
- Nonsterile
Certifications
Polypropylene plates are certified DNase- and RNase-free.
Specifications
Standard | |
8 x 12 | |
Applied Biosystems GeneAmp™ PCR System 9600b, Barnstead Thermolyne Amplitron II™, Techne™ Cyclogene, and Gene E with 96 x 0.2 mL block | |
96 | |
Round | |
25/Cs. | |
25/Cs. |
Without Lid | |
Clear | |
Polycarbonate | |
Non-sterile | |
PCR | |
1/Pk. | |
Clear |