Learn More
Corning™ Thermowell™ GOLD 384-Well Polypropylene PCR Microplates
GREENER_CHOICE
Fit automated equipment as well as the original manufacturers' plates
Brand:  Corning™ 3757
Product Code. 10651043
  • 684.84 € / Pack of 50
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Corning Thermowell GOLD 384-well PCR microplates feature exceptional dimensional stability following thermal cycling, and are fully compatible with automation, commonly used thermal cyclers, and Applied Biosystems™ sequencing adapters (see compatibility table).
Order Info
Packaged in packs of 10.
Certifications
Certified DNase- and RNase-free.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
16 x 24
Polypropylene
Non-sterile
For automation
10/Pk.
Clear
Without Lid
Clear
384
Round, Conical Bottom (V-well)
50/Cs.
50/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products