Learn More
Corning™ Thermowell™ Gold PCR Cap Strips
GREENER_CHOICE
Fit all Thermowell Gold 1 x 8-well PCR strips and 96-well PCR microplates
Brand:  Corning™ 3743
 View more versions of this product
Product Code. 10307722
  • 519.85 € / Pack of 1250
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Tube strips consist of eight 0.2 mL thin wall polypropylene tubes connected together. Dual connectors between adjacent tubes eliminate inadvertent breakage during sample handling. Tube strips are designed for precise fit in thermal cyclers to optimize heat transfer. Thermowell GOLD cap strips are sold separately from Thermowell GOLD tube strips. Original Thermowell tube strips and cap strips are packaged together. Tube strips are RNase-/DNase-free, and are autoclavable at 121°C.
Cat. No. 6547 has 60 of each color per bag; 1 bag of each color per case.
Cat. No. 3742 is designed for real-time PCR. Suitable for use with Thermowell GOLD 0.2 mL 1 x 8 PCR tube strips and 96-well microplates.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Non-sterile
1250/Cs.
PCR
Thermowell Gold 1 x 8-well PCR Strips and 96-well PCR Microplates
Polypropylene
Autoclavable
125/Pk.
Dome Cap Strip