Learn More
Corning™ Tubing Sets and Connectors for CellCube™
BIOPHARMAGREENER_CHOICE
Closed system cell cube, a new closed system offering designed to help reduce the risk of adventitious contamination
Brand:  Corning™ 3234
1035.50 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Product Code. 17229109
  • 1090.00 € / Pack of 4
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Closed System Cell Cube
  • Helps reduce the risk of adventitious contamination
  • CellCube 10, 25, 100 modules, circulation loops, and connectors
  • Allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik™ and MPC connectors.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Medium Filter, 2 Aseptic Connectors
4/Cs.
1/Pk.