missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Universal CellSTACK™ Caps
BIOPHARMAGREENER_CHOICE

For filling, venting, and manipulation

Brand:  Corning™ 3282

 View more versions of this product

Product Code. 10134212

  • 249.98 € / Pack of 5
Estimated Shipment: 29-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Choice of 33mm polyethylene, solid or vented
  • Sterilized by gamma irradiation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Orange
3.3 cm
With 1/8 in. (3.2mm) ID Tubing and a Female Luer 1/8 in. (3.2mm) Hose Barb with Male Luer Lock Plug
Single-use
Screw
HDPE
For filling, venting, and manipulation
Cap, Filling
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number17024003 Certificate TypeCertificate of Quality Date21/02/2025 Catalog Number3282
Lot Number17024003 Certificate TypeCertificate of Quality Date19/02/2025 Catalog Number3282
Lot Number07224002 Certificate TypeCertificate of Quality Date09/10/2024 Catalog Number3282
Lot Number07224001 Certificate TypeCertificate of Quality Date07/10/2024 Catalog Number3282
Lot Number07224003 Certificate TypeCertificate of Quality Date10/08/2024 Catalog Number3282
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Universal CellSTACK™ Caps > With 1/8 in. (3.2mm) ID Tubing and a Female Luer 1/8 in. (3.2mm) Hose Barb with Male Luer Lock Plug

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.