missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Universal Fit Pipet Tips: Bulk Packed
GREENER_CHOICE

Designed to assure a reliable fit with major brand pipetters

39.66 € - 411.00 €

Specifications

Sterility Non-sterile
Format Bulk
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
View More Specs

Products 6
Product Code Brand Format Volume (Metric) Color Price Quantity & Availability  
Product Code Brand Format Volume (Metric) Color Price Quantity & Availability  
10380331
GREENER_CHOICE
View Documents
Corning™
4844
Bulk 1 to 200 μL Natural
334.84 €
Pack of 10000
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
10299141
GREENER_CHOICE
View Documents
Corning™
4845
Bulk 1 to 200 μL Yellow
336.00 €
Pack of 10000
In Stock
to see stock.
Add to basket
 
10669321
GREENER_CHOICE
View Documents
Corning™
4846
Bulk 100 to 1000 μL Blue
411.00 €
Pack of 10000
In Stock
to see stock.
Add to basket
 
10005451
GREENER_CHOICE
View Documents
Corning™
4862
Bulk 1 to 200 μL Natural
39.66 €
Pack of 1000
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
10153761
GREENER_CHOICE
View Documents
Corning™
4866
Bulk 1 to 200 μL Yellow
39.66 €
Pack of 1000
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
10163761
GREENER_CHOICE
View Documents
Corning™
4868
Bulk 100 to 1000 μL Blue
41.70 €
Pack of 1000
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
Description

Description

Designed to assure a reliable fit with major brand pipetters

  • Beveled orifice helps ensure accurate fluid delivery
  • 1–200μL tips are graduated at 10, 50 and 100μL
  • 100–1000μL tips are graduated at 250, 500, and 1000μL
  • Choice of sterile or nonsterile
  • Bulk packed tips are nonsterile and very economical

Specifications

Specifications

Non-sterile
Universal Fit
Polypropylene
Bulk
Yes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.