Learn More
Falcon™ Vented Screw Caps for Flasks, Sterile
GREENER_CHOICE
Designed for use with Falcon 25-, 75-, and 175cm2 flasks
Brand:  Falcon™ 354639
 View more versions of this product
Product Code. 10709471
  • 102.00 € / Pack of 50
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ensure consistent gas exchange and minimize contamination
  • Polyethylene caps with an integral, hydrophobic 0.2 μm microporous membrane filter vent allow consistent gas exchange when the caps are in the closed position.
  • Gases required or cell growth and metabolism pass freely through the vent while microorganisms cannot. Falcon vented caps will not wet out.
  • Vented caps minimize contamination associated with standard open incubation. The vented caps prevent media that can become trapped in a partially opened cap from blocking gas exchange. The caps will not fall off in the incubator because of vibration.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyethylene
Falcon™ 175 cm2 flasks
Cap, Vented
Screw Cap
Blue
0.2μm Membrane vent
Sterile