Learn More
Corning™ Vitronectin, Human
Promotes cell attachment, spreading, proliferation, and differentiation of many normal and neoplastic cells; used to study cell migration
Brand: Corning™ 354238
This item is not returnable.
View return policy
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Specifications
Cell Attachment and Proliferation | |
Human Vitronectin | |
>90% by SDS-PAGE | |
Human Plasma | |
Human |
Lyophilized (dialyzed against 10 mm Phosphate Buffer pH 7.7); reconstitute in dH2O or Buffered solution at neutral pH | |
7.7 | |
0.25 mg | |
2° to 8°C | |
Lyophilized |
Product Content Correction
Product Title
Corning™ Vitronectin, Human > Vitronectin, Human; 0.25 mg
Spot an opportunity for improvement?Share a Content Correction