missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coronene, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 438°C to 440°C |
Color | Green to Yellow |
Boiling Point | 525°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% |
Packaging | Glass bottle |
Quantity | 100 mg |
Merck Index | 15, 2523 |
Solubility Information | Solubility in water: insoluble. Other solubilities: insoluble in concentrated sulfuric acid,slightly soluble in benzene |
Formula Weight | 300.35 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0455798 | Certificate of Analysis | 31/08/2023 | 110511000,110515000 |
A0395931 | Certificate of Analysis | 15/12/2022 | 110511000,110515000 |
A0414752 | Certificate of Analysis | 13/12/2022 | 110511000,110515000 |
A0438299 | Certificate of Analysis | 03/02/2022 | 110511000,110515000 |
A0424457 | Certificate of Analysis | 14/01/2021 | 110511000,110515000 |
Safety and Handling

Product Identifier
- Coronene
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Coronene, 95%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction