missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Stainless Steel NMR Spatula

Nickel Electro™ Stainless Steel NMR Spatula is recommended to be used for small powder samples.

Brand:  Nickel Electro™ 9576

Product Code. 12892326

  • 9.90 € / Each
Estimated Shipment: 14-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless steel
  • Non-magnetic
  • Bright finish
  • Recommended for small powder samples for introducing into 5mm OD NMR tubes.
  • The spatula is 250mm long allowing you to reach the very bottom of the tube to remove samples easily.
  • The rounded end of the spatula with curved tip can be used as a scoop whilst flat end can be used to break stuck samples.
  • Both ends are 3.5mm wide
Specifications

Specifications

1 Ea.
Spatula
Stainless Steel
PTFE
260°C
250 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers