Learn More
Isofield™ Cos Non-Sterile Lightweight Cleanroom Sock - GMP Annex 1 Compatible
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Size:
Large/X-Large
Small/Medium
Description
- Non-sterile lightweight
- Cleanroom socks
- Single-use
- 95% polyester, 5% spandex
- Mid-calf length
- Low linting
- White color
Specifications
Specifications
Size | Large/X-Large |
Product Type | Cleanroom Socks |