missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Color:White; Sterility:Sterile; Lid:With Lid

Brand:  Corning™ 3917

Product Code. 10167481

  • 700.00 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

 

  • Minimal well-to-well crosstalk
  • White microplates enhance luminecent signals and have low background luminescence
  • Recommended working volumes of 75 to 200μL
  • Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
  • Medical-grade polystyrene
  • Well volume: 320μL to 360μL
  • Working volume: 75μL to 200μL
  • Choice of flat, round or V-shaped bottom
  • Treated surface options are not coated, require no special handling and are stable at room temperature
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
  • Plate dimensions meet proposed industry standards

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
8 x 12
White
Polystyrene
75 to 200 μL
Cell Culture
Attachment and growth of anchorage-dependent cells
With Lid
20/Pk., 100/Cs.
Assay Plate
96
Sterile
Flat Bottom (F-well)
100/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.