Learn More
Corning™ Clear Polystyrene 96-Well Microplates
GREENER_CHOICE
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
Brand:  Corning™ 3369
344.30 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Product Code. 10731463
  • 362.42 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
High Binding
Without Lid
High Bind
Polystyrene
75 to 200 μL
High Binding
Capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions.
25/Pk.
Standard
8 x 12
Clear
96
Non-sterile
Flat Bottom (F-well)
100/Cs.
100/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Clear Polystyrene 96-Well Microplates > Standard; 25/Pk; No; High Binding; Easy Wash