missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well, High Binding, Flat-Bottom, Half-Area Microplate
Color:Clear; Sterility:Non-sterile; Lid:Without Lid
Brand: Corning™ 3690
This item is not returnable.
View return policy
Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Alphanumeric | |
Without Lid | |
High Bind | |
Plate readers | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. |
Half-Area | |
8 x 12 | |
Clear | |
Polystyrene | |
25 to 125 μL | |
High Binding | |
Ideal for immunoassays | |
25/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction