Learn More
Corning™ 96-Well, Non-Treated, Flat-Bottom, Half-Area Microplate
GREENER_CHOICE
Color:Black; Sterility:Non-sterile; Lid:Without Lid
Brand:  Corning™ 3694
Product Code. 10634102
  • 593.90 € / Pack of 100
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Alphanumeric
    Without Lid
    Black
    Polystyrene
    25 to 125 μL
    Non-treated
    Suitable for immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
    25/Pk.
    Half-Area
    8 x 12
    Plate readers
    96
    Non-sterile
    Flat Bottom (F-well)
    100/Cs.
    100/Cs.