Learn More
Corning™ 8-Channel Adapter for Vacuum Aspirator
For use with Corning Vacuum Aspirator. This 8-Channel Adapter can be used to remove media and solutions from 8 wells simultaneously on 96 well plate.
Brand:  Corning™ 4931
258.36 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Product Code. 10327781
  • 273.38 € / Each
Estimated Shipment: 02-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
This is an 8-channel adapter that fits on the Aspirator (No. 4930) so that it can be used to remove media and solutions from 8 wells simultaneously on a 96 well plate. 
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard 1-200μL disposable pipet tips
8
Corning Vacuum Aspirator (4930)
Not Autoclavable
Adapter, 8 channel