Learn More
Corning™ Clear Polystyrene 96-Well Microplates
GREENER_CHOICE
Product Code. 10367841 Shop All Corning Products
Click to view available options
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round
Surface Treatment:
Non-treated
Treated
Sterility:
Non-sterile
Sterile
Lid:
With Lid
Without Lid
Unit Size:
Pack of 100
Pack of 48
Pack of 50
Product Code. 10367841
Brand: Corning™ 3896
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Not Treated
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
For Use With (Application) Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
No. per Case 48/Cs.
No. per Pack 1/Pk.
Quantity 48/Cs.