missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Cluster Tube System
Manufactured from polypropylene for strength and durability. Corning™ Cluster Tube System is ideal for harvesting cells, cell uptake and receptor binding assays, and radioimmunoassays.
Brand: Corning™ 4408
125.67 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Description
- Composed of 96 polypropylene tubes in a standard microplate format.
- Features 1.2 mL tubes that are available individually or in strips of eight tubes.
- Polyethylene tube caps are available in 8-cap strips.
Specifications
Strip | |
Autoclavable | |
8-Strip Microplate Cluster Tube | |
120/Pk. | |
Translucent | |
Standard 96-well microplate for harvesting cells, cell uptake and receptor binding assays and radioimmunoassays, useful with single and multichannel pipetters and in robotic workstations and centrifuges | |
96 |
Bulk, 120/Bag | |
Polypropylene | |
Non-sterile | |
120/Cs. | |
1 mL | |
0.033 oz. |