missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Costar™ Disposable Traditional Style Polystyrene Storage Bottles
Manufactured from clear polystyrene for strength and optimum visibility. Corning™ Costar™ Disposable Storage Bottles offer the ideal solution for storing media, buffers, and other aqueous solutions
77.32 € - 112.76 €
Specifications
Color | Clear |
---|---|
Material | Polystyrene |
No. per Pack | 1/Pk. |
Sterility | Sterile |
Closure Color | Red |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10052491
|
Corning™
8388 |
125 mL |
98.10 €
Pack of 24 |
In Stock
Log in to see stock. |
|||||
10539082
|
Corning™
8390 |
250 mL |
87.64 €
Pack of 12 |
In Stock
Log in to see stock. |
|||||
10179200
|
Corning™
8393 |
500 mL |
77.32 €
Pack of 12 |
|
|||||
10177500
|
Corning™
8396 |
1 L |
112.76 €
Pack of 12 |
|
|||||
Description
- Disposable polystyrene bottles for storage of media, buffers, and other aqueous solutions
- Costar™ traditional style has smooth sides
- Plug seal caps (45 mm) provide an airtight seal and help minimize the risk of contamination.
- Bottles can be used with Corning vacuum filtration systems with 45 mm neck sizes (Cat. Nos. 430626, 430627, 431161, 430049, 430513, 430514,431118, 430015, and 431174).
- Sterile
- Nonpyrogenic
Specifications
Clear | |
1/Pk. | |
Red | |
Cylindrical | |
45 mm |
Polystyrene | |
Sterile | |
45 | |
HDPE | |
Plug Seal |