missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Costar™ Transwell™-COL Collagen-Coated PTFE Membrane Insert
Shop All Corning Products

Click to view available options
Pore Size:
0.4 μm
3 μm
For Use With (Equipment):
12 Well Plate
24 Well Plate
6 Well Plate
Unit Size:
Pack of 24
Description
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
You can use cell culture inserts to:
- Produce a cell culture environment that closely resembles an in vivo state
- Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
- Co-culture cells with or without cell-to-cell contact
- Design a diversity of experiments using various pore sizes, membrane types, and coatings Transparent, Collagen-coated PTFE membrane
- Promotes cell attachment and spreading
- Equimolar mixture of Collagen type I and III
- Individually packaged
- Multiple well plates included in each case
- Sterile
Order Info
Individually packed and supplied in package with tissue culture plates with lids; sterile.
Specifications
Specifications
| Culture Area | 1.12 cm2 |
| Material | PTFE |
| Volume (Metric) Well | 1.5 mL |
| Pore Size | 3 μm |
| Type | Membrane Insert |
| For Use With (Equipment) | 12 Well Plate |
| Diameter (Metric) Filter | 12 mm |
| No. per Pack | 1/Pk. |
| No. per Case | 24/Cs. |
| Surface Coating | Collagen |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction