missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Cotton Cord

Prolongs delivery time and aids in measurement accuracy. BRAND™ Cotton Cord is made of 100% cotton wool.

Brand:  Brand™ 28205

108.60 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10473521

  • 126.00 € / 1kg
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Cotton plugs may prolong delivery time and may therefore influence the accuracy of measurement. In tests using pipettes that have had cotton plugs inserted by a cotton plugging machine, the results were within the indicated error limits.
  • 100% Cotton wool
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cotton Wool
Pipets
13 g/10 m
Cotton Wool
1 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.