missing translation for 'onlineSavingsMsg'
Learn More

Cottonseed oil, pure, Thermo Scientific Chemicals

Product Code. 10552391
Click to view available options
Quantity:
500 mL
2.5 L

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-29-4
MDL Number MFCD00130872

Specifications

Acidity 0.5mg KOH/g max.
Color Yellow
Density 0.9100g/mL
Flash Point >112°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4720 to 1.4730
Quantity 500 mL
Saponification Value 185 to 198mg KOH/g
Merck Index 10,2535
Specific Gravity 0.91
Solubility Information Solubility in water: .. Other solubilities: soluble in ether,benzene,chloroform and carbon,disulfide,slightly soluble in alcohol
Grade Pure
Physical Form Oil
Chemical Name or Material Cottonseed oil
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.