Learn More
Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
59.30 € - 184.92 €
Chemical Identifiers
CAS | 531-81-7 |
---|---|
Molecular Formula | C10H6O4 |
Molecular Weight (g/mol) | 190.154 |
MDL Number | MFCD00006852 |
InChI Key | ACMLKANOGIVEPB-UHFFFAOYSA-N |
Synonym | coumarin-3-carboxylic acid, 2-oxo-2h-chromene-3-carboxylic acid, 3-carboxycoumarin, 2-oxobenzopyran-3-carboxylic acid, g 1 rodenticide, 2h-1-benzopyran-3-carboxylic acid, 2-oxo, 2-oxo-2h-1-benzopyran-3-carboxylic acid, unii-v85uov8788, g 1 the rodenticide van, g 1 van |
PubChem CID | 10752 |
IUPAC Name | 2-oxochromene-3-carboxylic acid |
SMILES | C1=CC=C2C(=C1)C=C(C(=O)O2)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11327156
|
Thermo Scientific Alfa Aesar
L07133.14 |
25 g |
59.30 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337156
|
Thermo Scientific Alfa Aesar
L07133.22 |
100 g |
184.92 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Coumarin-3-carboxylic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCoumarin-3-carboxylic acid is used as pharmaceutical intermediate.
Solubility
Soluble in water (13 g/L at 37°C) and alcohol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
531-81-7 | |
190.154 | |
ACMLKANOGIVEPB-UHFFFAOYSA-N | |
10752 | |
C1=CC=C2C(=C1)C=C(C(=O)O2)C(=O)O |
C10H6O4 | |
MFCD00006852 | |
coumarin-3-carboxylic acid, 2-oxo-2h-chromene-3-carboxylic acid, 3-carboxycoumarin, 2-oxobenzopyran-3-carboxylic acid, g 1 rodenticide, 2h-1-benzopyran-3-carboxylic acid, 2-oxo, 2-oxo-2h-1-benzopyran-3-carboxylic acid, unii-v85uov8788, g 1 the rodenticide van, g 1 van | |
2-oxochromene-3-carboxylic acid |
Specifications
531-81-7 | |
C10H6O4 | |
UN2811 | |
14,2563 | |
Soluble in water (13g/L at 37°C) and alcohol. | |
C1=CC=C2C(=C1)C=C(C(=O)O2)C(=O)O | |
190.154 | |
190.15 | |
Coumarin-3-carboxylic acid |
190°C to 193°C | |
MFCD00006852 | |
154276 | |
coumarin-3-carboxylic acid, 2-oxo-2h-chromene-3-carboxylic acid, 3-carboxycoumarin, 2-oxobenzopyran-3-carboxylic acid, g 1 rodenticide, 2h-1-benzopyran-3-carboxylic acid, 2-oxo, 2-oxo-2h-1-benzopyran-3-carboxylic acid, unii-v85uov8788, g 1 the rodenticide van, g 1 van | |
ACMLKANOGIVEPB-UHFFFAOYSA-N | |
2-oxochromene-3-carboxylic acid | |
10752 | |
98% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 208-518-0
missing translation for 'rtecsNumber' : DJ2490000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- Coumarin-3-carboxylic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
- MIXTURE LIST-Contains : 2-Oxo-2H-chromene-3-carboxylic acid
RUO – Research Use Only