missing translation for 'onlineSavingsMsg'
Learn More

Coumarin-6-sulfonyl chloride, 97%, Thermo Scientific Chemicals

57.20 € - 655.00 €

Chemical Identifiers

CAS 10543-42-7
Molecular Formula C9H5ClO4S
Molecular Weight (g/mol) 244.65
MDL Number MFCD01941320
InChI Key HQIPMBGUDSOVEA-UHFFFAOYSA-N
Synonym 2-oxo-2h-chromene-6-sulfonyl chloride, coumarin-6-sulfonyl chloride, 6-cs-cl, coumarin-6-sulphonyl chloride, 2-oxo-2h-chromene-6-sulphonyl chloride, 2-oxo-2h-1-benzopyran-6-sulfonyl chloride, 6-chlorosulfonyl chromen-2-one, pubchem5506, c6scl, timtec-bb sbb002796
PubChem CID 2735833
IUPAC Name 2-oxochromene-6-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C2OC(=O)C=CC2=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15470968
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H54767.03
1 g
57.20 €
1g
Please to purchase this item. Need a web account? Register with us today!
15480968
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H54767.06
5 g
182.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
15490968
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H54767.14
25 g
655.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Coumarin-6-sulfonyl chloride is used as a derivatizing agent for amines and phenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Coumarin-6-sulfonyl chloride is used as a derivatizing agent for amines and phenols.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
Specifications

Chemical Identifiers

10543-42-7
244.65
HQIPMBGUDSOVEA-UHFFFAOYSA-N
2735833
ClS(=O)(=O)C1=CC=C2OC(=O)C=CC2=C1
C9H5ClO4S
MFCD01941320
2-oxo-2h-chromene-6-sulfonyl chloride, coumarin-6-sulfonyl chloride, 6-cs-cl, coumarin-6-sulphonyl chloride, 2-oxo-2h-chromene-6-sulphonyl chloride, 2-oxo-2h-1-benzopyran-6-sulfonyl chloride, 6-chlorosulfonyl chromen-2-one, pubchem5506, c6scl, timtec-bb sbb002796
2-oxochromene-6-sulfonyl chloride

Specifications

10543-42-7
MFCD01941320
196167
2-oxo-2h-chromene-6-sulfonyl chloride, coumarin-6-sulfonyl chloride, 6-cs-cl, coumarin-6-sulphonyl chloride, 2-oxo-2h-chromene-6-sulphonyl chloride, 2-oxo-2h-1-benzopyran-6-sulfonyl chloride, 6-chlorosulfonyl chromen-2-one, pubchem5506, c6scl, timtec-bb sbb002796
HQIPMBGUDSOVEA-UHFFFAOYSA-N
2-oxochromene-6-sulfonyl chloride
2735833
97%
C9H5ClO4S
UN3261
Moisture sensitive
Slightly soluble in water.
ClS(=O)(=O)C1=CC=C2OC(=O)C=CC2=C1
244.65
244.65
Coumarin-6-sulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only