missing translation for 'onlineSavingsMsg'
Learn More

Coumarin, 98%, Thermo Scientific Chemicals

Product Code. 11438246
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Coumarin is an anticoagulant found in medicinal plants and used in phytomedicine for the treatment of venous diseases. This compound is found to have anti-tumor activities in several human cells lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-64-5
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.15
MDL Number MFCD00006850
InChI Key ZYGHJZDHTFUPRJ-UHFFFAOYSA-N
Synonym coumarin, 2h-chromen-2-one, 2h-1-benzopyran-2-one, cumarin, 1,2-benzopyrone, rattex, tonka bean camphor, coumarinic anhydride, coumarine, benzo-alpha-pyrone
PubChem CID 323
ChEBI CHEBI:28794
SMILES O=C1OC2=CC=CC=C2C=C1

Specifications

Melting Point 68°C to 71°C
Density 0.935
Boiling Point 297°C to 299°C
Flash Point 162°C (323°F)
Odor Pleasant
Quantity 1000 g
UN Number UN2811
Beilstein 383644
Merck Index 14,2562
Solubility Information Soluble in water.
IUPAC Name chromen-2-one
Formula Weight 146.15
Percent Purity 98%
Chemical Name or Material Coumarin
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.