missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ CountBright™ Absolute Counting Beads, for flow cytometry

CountBright™ Absolute Counting Beads, for flow cytometry

Brand:  Molecular Probes™ C36950

403.75 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 11570066

  • 425.00 € / 5mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

CountBright™ absolute counting beads are a calibrated suspension of microspheres that are brightly fluorescent across a wide range of excitation and emission wavelengths (UV to 635 nm excitation and 385-800 nm emission). CountBright™ absolute counting beads are mixed with the cell sample and assayed via flow cytometry. By comparing the ratio of bead events to cell events, absolute numbers of cells in the sample can be calculated. Because CountBright™ beads are mixed in the test sample, absolute cell counts using this single-platform method are more accurate and less complicated than cell concentration determined using multiple-platform testing. CountBright™ absolute counting beads can be used with any sample type, including no-wash/lysed whole blood.

View information about all flow cytometry cell counting beads.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains 1 bottle of CountBright™ absolute counting beads (5 mL). Store in refrigerator (2°C to 8°C) and protect from light.
7 μm
UV to 635/385 to 800 nm
Solution
5 mL
CountBright™
Fluorescence
any lasers (UV to 633/635)
Flow Cytometer
100
Room Temperature
Cell Counting Bead
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.