missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CountBright™ Plus Absolute Counting Beads

Calibrated suspension of ∼4 μm beads that encapsulate dyes that are excitable across a wide range of UV-IR wavelengths and brightly fluoresce in the emission range of 385-860 nm.

Brand:  Invitrogen™ C36995

336.30 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.


Product Code. 16498535

  • 354.00 € / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Help to determine the count.
  • The concentration of cells in the original sample.
  • Beads are on-scale with most cell types, including human whole lysed blood, for ease of use
  • >95% singlets for improved accuracy
  • Excited by UV-IR lasers for an expanded emission range (385–860 nm)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

  • 5 mL beads in buffer
  • Store at 2°C to 8°C
100 tests
CountBright™
4 μm
Room Temperature
Cell Counting Bead
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.