Learn More
Reichert Instruments™ Counter Push Button
For use with the Darfield Quebec colony counter
Brand:  Reichert Instruments™ 13332400
Product Code. 12782585
  • 264.00 € / Each
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Darkfield Quebec colony counter allows the user to quickly and accurately count bacteria colonies.
It is available in two models for digital or manual counting. Both models provide even, glare-free illumination as light is spread uniformly over the entire culture plate.
Adjustable dish holder for centering both round dishes and square culture plates
  • Adjustable focusing rod
  • Lens rotates a full 360°
  • Built-in tilt leg
  • Optional 1.5x auxiliary lens fits over standard lens, increasing magnification to 3x
Specifications
Specifications
Counter Push Button
1Each
For Use with Quebec™ Darkfield Digital Colony Counters