Learn More
Thermo Scientific™ Countess™ Disposable Slide Holder
For use with Countess™ II FL Automated Cell Counter Only.
Brand:  Thermo Scientific™ AMEP4745
Additional Details : Weight : 0.70000kg
Product Code. 15594183
  • 28.03 € / Each
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
Description
Description
For use with Countess™ II FL Automated Cell Counter Only. Holds Countess™ disposable slides. Not compatible with the Countess™ II FL Reusable Slide (A25750).
Specifications
Specifications
Disposable Slide Holder
Cell Counting
Room Temperature
Countess™
Originally supplied with CIIFL Automated Counter