Learn More
Corning™ Counting Chamber, 0.2 mm
Reusable glass cell counting slide for use with the Corning™ Cell Counter. This gridless slide has a 0.1 or 0.2 mm depth and comes standard with two high quality cover slips. These slides have a notch for easy loading of cell suspension.
Brand:  Corning™ 480201
Product Code. 17797014
  • 129.21 € / Each
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made with reusable glass
  • Gridless
  • 0.2 mm depth
  • Includes two high quality cover slips
  • Feature a notch for easy loading of cell suspension
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Counting Chamber
20 μL
Corning™ Cell Counter
Glass
0.2 mm
1 Pk.