missing translation for 'onlineSavingsMsg'
Learn More

Marienfeld Superior™ Counting Chamber with Bright Line Double Net Ruling

Marienfeld Superior™ Counting Chamber with double net ruling are precise measuring instruments for investigation of cell suspensions

Brand:  Marienfeld Superior™ 0640030

 View more versions of this product

Product Code. 13444890

  • 362.00 € / Each
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Precise measuring instrument for determination of the number of particles in liquids
  • Made of special optical glass
  • Equipped with two grids
  • With 2 haemacytometer cover glasses 20x26 mm
  • Individually packed in a transparent plastic box, 10 boxes in a carton
Specifications

Specifications

Counting chamber, bright line
CE-marked (IVD)
Special optical glass
0.1 mm
Neubauer Improved
0.0025 mm2
1
Two cover glasses 20 x 26 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.