Learn More
Hirschmann™ Counting Chambers
Facilitates quick and easy determination of cell concentration in liquid samples. Hirschmann Instruments™ Counting Chambers are available in a variety of depths to fit a range of systems.
Brand:  Hirschmann™ 8110204
 View more versions of this product
Product Code. 10527831
  • 174.00 € / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Double net ruling
  • Available bright-lined or no, with or without clamps
  • Includes 2 cover glasses for haemocytometer
  • CE acc. IVD 98/79 EC
Specifications
Specifications
Counting Chambers, Bright lined with clamps; Neubauer-improved
1Each
0.1mm
2 Haemocytometer Glass Covers and Clamps