Learn More
Hirschmann™ Counting Chambers
Facilitates quick and easy determination of cell concentration in liquid samples. Hirschmann™ Counting Chambers are available in a variety of depths to fit a range of systems.
Brand:  Hirschmann™ 8110111
 View more versions of this product
Product Code. 10603972
  • 222.00 € / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Double net ruling
  • Including 2 cover glasses for hemacytometer
  • CE acc. IVD 98/79 EC
Specifications
Specifications
Counting chambers, double net ruling, with clamps
1 Each
CE acc. IVD 98/79 EC
Counting chamber acc. to Nageotte
0.5 mm
2 cover glasses for haemacytometer