Learn More
Hirschmann™ Counting Chambers with Clamps
With double clamps for secure positioning.
Brand: Hirschmann™ 8110103
This item is not returnable.
View return policy
Description
- Double net ruling
- With clamps
- Includes 2 haemocytometer glass covers
- With LOT number on packaging
- Labelled with: usable until, CE pursuant to IVD 98/79 EC
Specifications
Counting Chamber (Neubauer) | |
0.1 mm | |
2 haemocytometer glass covers |
Double net ruling | |
CE pursuant to IVD 98/79 EC |