missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Cover

A 13 Lid for fixing delicate wash items

Brand:  Miele Professional™ 3810200

Product Code. 10724333

  • 76.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Lid in stainless steel
  • 1 mm wire, 8 mm mesh size, 4 mm frames
Specifications

Specifications

Stainless Steel
Stainless Steel
20 mm
Cover
320 mm
200 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers