Learn More
Karl Hecht™ Assistent™ Cover Glasses for Hemocytometers
Prevents color distortion due to super-transparent and colorless construction. Karl Hecht™ Assistent™ Cover Glasses for Hemocytometers have a refractive index.
Brand:  Karl Hecht™ 40415710
Product Code. 10647293
  • 19.00 € / Pack of 10
Estimated Shipment: 28-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Excellent flatness
  • High spectral transmission
  • Optimum resistance to chemical attack
  • Low levels of inclusions, striae, bubbles, streaks, etc.
Specifications
Specifications
Cover Glass
0.4 mm
For Hemacytometer