Learn More
DuPont™ Tyvek 200 Easysafe Hooded Coverall Product Code.: 15360541
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
DuPont™ Tyvek 200 Easysafe Hooded Coverall
GREENER_CHOICE
Product Code. 15360541 Shop All DuPont Products
3X-Large
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Product Code. 15360541
Brand: DuPont™ D14886081
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
Add to basket
Offering adequate Type 5 and 6 barrier protection and remains both air and moisture vapour permeable. DuPont™ Tyvek™ 200 Easysafe Hooded Coveralls are made from a new and unique optimized polyethylene nonwoven that is softer with a more open structure than Tyvek™ 500 and 600.
  • Robust yet lightweight
  • Elasticated face, wrists, waist and ankles
  • Antistatic treatment on the outside
  • Stitched external seams in yellow for visual identification and differentiation
Greener Choice Claims
  • Environmental benefits include:
    • Recycled Content
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls
Size 3X-Large
Material Polyethylene
Color White
Ankle Style Elastic
Neck Style Hooded
Hood Type 2-panel
Closure Type Zipper
Seam Type Stitched
Protection Class Category III, Type 5 and 6
Certifications/Compliance (EU) 2016/425, EN 1073-2, EN 1149-1
Size (Metric) Chest 124 to 132 cm
Product Title
DuPont™ Tyvek 200 Easysafe Hooded Coverall >