Learn More
COXIV Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11242-1-AP-150UL
Product Code. 16868183
  • 189.00 € / 150µL
Explore more special offers
Description
Description
This antibody was generated against full length COX4I1 protein and cross reacts with COX4I2.
Cytochrome c oxidase (COX) is the terminal enzyme of the mitochondrial respiratory chain. It is a multi-subunit enzyme complex that couples the transfer of electrons from cytochrome c to molecular oxygen and contributes to a proton electrochemical gradient across the inner mitochondrial membrane. The complex consists of 13 mitochondrial- and nuclear-encoded subunits. The functions of the nuclear-encoded subunits are unknown but they may play a role in the regulation and assembly of the complex. COX4 (COX4I1) is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
Specifications
Specifications
COXIV
Polyclonal
Unconjugated
COX4I1
COX IV 1, COX4, COX4I1, COXIV, Cytochrome c oxidase polypeptide IV, Cytochrome c oxidase subunit 4 isoform 1 mitochondrial
Rabbit
Antigen Affinity Chromatography
RUO
12857, 1327, 29445
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P10888, P13073, P19783
COX4I1
COXIV Fusion Protein Ag1640
150 μL
Primary
Human, Mouse, Rat