Learn More
Cole-Parmer™ CPC™ Panel Mount Gasket Couplers
Virtually eliminate any loss of product with this leak-free design
Brand:  Cole-Parmer™ 31603-18
 View more versions of this product
Product Code. 15427333
  • 50.10 € / Pack of 10
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance.
  • For a complete fitting, order one body and one insert.
  • All bodies will mate with inserts to make a fitting that will work for multiple applications.
Specifications
Specifications
Panel Mount Gasket
10 Gaskets
NS4 Series Couplers
EPDM