Learn More
Cole-Parmer™ CPC™ Polysulfone Coupling Cap
Features a smooth inner bore and leak-free seal protect against contamination
Brand:  Cole-Parmer™ 31310-02
Product Code. 15437153
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • With their locking feature, these couplings guard against accidental disconnect and spills.
  • To assemble, place the insert in the coupling body and twist to lock.
  • An audible click ensures your connection.
  • All couplings are made from USP Class VI-compliant polysulfone with platinum-cured silicone seals.
  • Available in 3/4in. size, these fittings allow quick fill or empty of bioprocessing bags.
Specifications
Specifications
Coupling Cap, Polysulfone
10/Pk.
Autoclave, Ethylene Oxide (ETO) or Gamma
3/4 in.
Quick-Disconnect x Barb
Polysulfone