missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ CPC™ Quick-Disconnect Valved Coupling Body

Quick-disconnect fitting with simple push-to-connect interface for your rigid tubing

Brand:  Cole-Parmer™ 31304-02

 View more versions of this product

Product Code. 15497093

  • 24.85 € / Each
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Plug insert into body to connect fitting.
  • Fitting halves lock in place immediately.
  • To disconnect fitting halves, simply press the thumb latch on the coupling body.
  • spring-operated valve in each fitting half seals the flow path.
  • Straight-through fittings without shutoff valves are also available.
  • All fitting bodies have 316 stainless steel springs and latches.
Specifications

Specifications

Fitting
1810°F
3/8 in.
Autoclave or Ethylene Oxide (ETO)
120 psi
White
82°C
Quick-Disconnect x Push-to-Connect
Acetal
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers