missing translation for 'onlineSavingsMsg'
Learn More

CPCCOEt, Tocris Bioscience™

Product Code. 30001164
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective non-competitive mGlu1 antagonist

hmGlu1 subtype-selective non-competitive antagonist (IC50 = 6.5 μM). Has no agonist or antagonist activity at hmGlu2, 4a, 5a, 7b, 8a or ionotropic receptors at concentrations of up to 100 μM.

Chemical Identifiers

CAS 179067-99-3
Molecular Formula C13H13NO4
Molecular Weight (g/mol) 247.25
InChI Key FXCTZFMSAHZQTR-DRYKPYBASA-N
Synonym cpccoet
PubChem CID 92460002
IUPAC Name ethyl (1aS,7Z,7aS)-7-hydroxyimino-1,7a-dihydrocyclopropa[b]chromene-1a-carboxylate
SMILES CCOC(=O)C12CC1C(=NO)C3=CC=CC=C3O2

Specifications

Quantity 10 mg
Formula Weight 247.25
Percent Purity >99%
Chemical Name or Material CPCCOEt
missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'