missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ CPE Disposable Shoe Cover (Universal Size)
Disposable spunbond polypropylene shoe cover, with anti-skid sole. Wear over your shoes in non-hazardous environments to protect against dirt and grime. Ideal for non-critical areas or pre-gowning entry rooms.
Brand: Fisherbrand™ FBHLSCCPE
This item is not returnable.
View return policy
Description
General purpose disposable shoe covers protect floors, footwear and promotes general hygiene.
- With elasticated tops for an easy universal fit
- High quality CPE (polythene) construction
- For use in the laboratory or wider working environment
- ISO tested to ISO3801 and ISO13934-1
- Supplied as a carton of 20 bags, 100 covers per bag
Specifications
Universal | |
Blue | |
Non-sterile | |
Plain | |
General Purpose in the Lab or Cleanroom | |
2000 Pk. | |
15 cm ±2 cm (Heel Height) |
CPE Polyethylene | |
Yes | |
Elasticated Top | |
22 μm | |
PPE CAT I, ISO Tested to ISO3801 and ISO13934-1 | |
38 ±2 cm Stretch Length | |
2.6 ±0.2 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ CPE Disposable Shoe Cover (Universal Size) > CPE Disposable Shoe Cover; Certifications/Compliance: PPE CAT I, ISO Tested to ISO3801 and ISO13934-1; Thickness: 22 μm; Sole Type: Plain; Size: Universal; Color: Blue; Closure Type: Elasticated Top
Spot an opportunity for improvement?Share a Content Correction